ContactUsLinkComponent
Mouse RANTES ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse RANTES in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 l of sample vol 1 * 96 Tests
Catalog # ANTIA323194-96
Supplier:  ANTIBODIES.COM
New Product
Mouse RANTES ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse RANTES in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 l of sample vol 1 * 96 Tests
Catalog # ANTIA323194-96
Supplier:  ANTIBODIES.COM