ContactUsLinkComponent
Beta Lactam + Cephalexin + Tetracycline Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determ ination of Beta Lactam + Cephalexin + Tetracycline in milk. 1 * 96 Tests
Catalog # ANTIA327182-96
Supplier:  ANTIBODIES.COM
New Product
Beta Lactam + Cephalexin + Tetracycline Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determ ination of Beta Lactam + Cephalexin + Tetracycline in milk. 1 * 96 Tests
Catalog # ANTIA327182-96
Supplier:  ANTIBODIES.COM