ContactUsLinkComponent
Horse IL-18 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of horse IL-18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4234-48
Supplier:  ANTIBODIES.COM
New Product
Horse IL-18 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of horse IL-18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4234-48
Supplier:  ANTIBODIES.COM