ContactUsLinkComponent
Rat Creatine Kinase MM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of rat Creatine Kinase MM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 48 Tests
Catalog # ANTIA3446-48
Supplier:  ANTIBODIES.COM
New Product
Rat Creatine Kinase MM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of rat Creatine Kinase MM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 48 Tests
Catalog # ANTIA3446-48
Supplier:  ANTIBODIES.COM