ContactUsLinkComponent
Human Bile Salt-Activated Lipase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Bile Salt-Activated Lipase in tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA5789-48
Supplier:  ANTIBODIES.COM
New Product
Human Bile Salt-Activated Lipase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Bile Salt-Activated Lipase in tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA5789-48
Supplier:  ANTIBODIES.COM