Porcine IL-15 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of porcine IL-15 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 96 Tests
Catalog # ANTIA4174-96
Supplier: ANTIBODIES.COM