ContactUsLinkComponent
Human Cadherin 16 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human Cadherin 16 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA3792-48
Supplier:  ANTIBODIES.COM
New Product
Human Cadherin 16 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human Cadherin 16 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA3792-48
Supplier:  ANTIBODIES.COM