Bovine TGF beta 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of bovine TGF beta 1 in serum, platelet-poor plasma, tissue homogenates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA4053-48
Supplier: ANTIBODIES.COM