ContactUsLinkComponent
Mouse Mannan Binding Lectin/MBL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Mannan Binding Lectin/MBL in serum, plasma, tissue homogenates, cell lysates, cell cultu 1 * 48 Tests
Catalog # ANTIA3776-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Mannan Binding Lectin/MBL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Mannan Binding Lectin/MBL in serum, plasma, tissue homogenates, cell lysates, cell cultu 1 * 48 Tests
Catalog # ANTIA3776-48
Supplier:  ANTIBODIES.COM