ContactUsLinkComponent
Guinea Pig RANTES ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of guinea pig RANTES in serum, plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 48 Tests
Référence ANTIA3991-48
Fournisseur:  ANTIBODIES.COM
New Product
Guinea Pig RANTES ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of guinea pig RANTES in serum, plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 48 Tests
Référence ANTIA3991-48
Fournisseur:  ANTIBODIES.COM