ContactUsLinkComponent
Kanamycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Kanamycin in raw co mmingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327220-96
Supplier:  ANTIBODIES.COM
New Product
Kanamycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Kanamycin in raw co mmingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327220-96
Supplier:  ANTIBODIES.COM