ContactUsLinkComponent
Human Mast Cell Chymase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human Mast Cell Chymase in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4834-48
Supplier:  Antibodies.com
New Product
Human Mast Cell Chymase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human Mast Cell Chymase in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4834-48
Supplier:  Antibodies.com