ContactUsLinkComponent
Thiamphenicol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Thiamphenicol i n raw commingled cow&#0146 s milk and processed milk products. 1 * 96 Tests
Catalog # ANTIA327259-96
Supplier:  ANTIBODIES.COM
New Product
Thiamphenicol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Thiamphenicol i n raw commingled cow&#0146 s milk and processed milk products. 1 * 96 Tests
Catalog # ANTIA327259-96
Supplier:  ANTIBODIES.COM