ContactUsLinkComponent
Porcine Fas Ligand ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of porcine Fas Ligand in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 96 Tests
Catalog # ANTIA3779-96
Supplier:  ANTIBODIES.COM
New Product
Porcine Fas Ligand ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of porcine Fas Ligand in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 96 Tests
Catalog # ANTIA3779-96
Supplier:  ANTIBODIES.COM