ContactUsLinkComponent
Mouse IL-1 Receptor I/IL-1R-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of mouse IL-1 Receptor I/IL-1R-1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3480-96
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-1 Receptor I/IL-1R-1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of mouse IL-1 Receptor I/IL-1R-1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3480-96
Supplier:  ANTIBODIES.COM