ContactUsLinkComponent
Mouse IL-17C ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IL-17C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 96 Tests
Catalog # ANTIA5011-96
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-17C ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IL-17C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 96 Tests
Catalog # ANTIA5011-96
Supplier:  ANTIBODIES.COM