ContactUsLinkComponent
Human Collagen IV alpha 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Collagen IV alpha 1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Katalognr. ANTIA4138-48
Supplier:  ANTIBODIES.COM
New Product
Human Collagen IV alpha 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Collagen IV alpha 1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Katalognr. ANTIA4138-48
Supplier:  ANTIBODIES.COM