ContactUsLinkComponent
Monkey TGF beta 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of monkey TGF beta 1 in serum, platelet-poor plasma, tissue homogenates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA4057-48
Supplier:  ANTIBODIES.COM
New Product
Monkey TGF beta 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of monkey TGF beta 1 in serum, platelet-poor plasma, tissue homogenates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA4057-48
Supplier:  ANTIBODIES.COM