ContactUsLinkComponent
Human Carbonic Anhydrase 3/CA3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human Carbonic Anhydrase 3/CA3 in serum, plasma, tissue homogenates, cell lysates, cell culture 1 * 96 Tests
  ANTIA5760-96
New Product
Human Carbonic Anhydrase 3/CA3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human Carbonic Anhydrase 3/CA3 in serum, plasma, tissue homogenates, cell lysates, cell culture 1 * 96 Tests
  ANTIA5760-96