ContactUsLinkComponent
Human IL-22 RA2/IL-22BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human IL-22 RA2/IL-22BP in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5426-48
Supplier:  ANTIBODIES.COM
New Product
Human IL-22 RA2/IL-22BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human IL-22 RA2/IL-22BP in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5426-48
Supplier:  ANTIBODIES.COM