Chloramphenicol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Chloramphenic ol in raw commingled cow’ s milk. 1 * 96 Tests
Catalog # ANTIA327189-96
Anbieter: ANTIBODIES.COM