ContactUsLinkComponent
Erythromycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Erythromycin in raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327203-96
Supplier:  ANTIBODIES.COM
New Product
Erythromycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Erythromycin in raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327203-96
Supplier:  ANTIBODIES.COM