ContactUsLinkComponent
Human Inhibin beta E Chain ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Inhibin beta E Chain in serum, plasma, tissue homogenates, cell lysates, cell culture superna 1 * 96 Tests
  ANTIA5246-96
New Product
Human Inhibin beta E Chain ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Inhibin beta E Chain in serum, plasma, tissue homogenates, cell lysates, cell culture superna 1 * 96 Tests
  ANTIA5246-96