Ractopamine + Clenbuterol Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Rac topamine + Clenbuterol in urine. 1 * 40 Tests
Catalog # ANTIA327243-40
Supplier: Antibodies.com