ContactUsLinkComponent
Human ATP-binding Cassette Sub-family A Member 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ATP-binding Cassette Sub-family A Member 3 in tissue homogenates or oth 1 * 96 Tests
Catalog # ANTIA5767-96
Supplier:  Antibodies.com
New Product
Human ATP-binding Cassette Sub-family A Member 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ATP-binding Cassette Sub-family A Member 3 in tissue homogenates or oth 1 * 96 Tests
Catalog # ANTIA5767-96
Supplier:  Antibodies.com