ContactUsLinkComponent
This kit uses click chemistry to conjugate four fluorochromes to primary antibodies after biotinylation. 1 * 2.000 µG
Catalog # ANTIA329038-4500
Supplier:  ANTIBODIES.COM
New Product
This kit uses click chemistry to conjugate four fluorochromes to primary antibodies after biotinylation. 1 * 2.000 µG
Catalog # ANTIA329038-4500
Supplier:  ANTIBODIES.COM