ContactUsLinkComponent
Human Complement C9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Complement C9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other b 1 * 96 Tests
Codice catalogo ANTIA5057-96
Nome del fornitore:  ANTIBODIES.COM
New Product
Human Complement C9 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Complement C9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other b 1 * 96 Tests
Codice catalogo ANTIA5057-96
Nome del fornitore:  ANTIBODIES.COM