ContactUsLinkComponent
Human PI 3 Kinase p110 delta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of human PI 3 Kinase p110 delta in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5423-96
Supplier:  ANTIBODIES.COM
New Product
Human PI 3 Kinase p110 delta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of human PI 3 Kinase p110 delta in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5423-96
Supplier:  ANTIBODIES.COM