ContactUsLinkComponent
Rat alpha Skeletal Muscle Actin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of rat alpha Skeletal Muscle Actin in tissue homogenates, cell lysates, cell culture supernates o 1 * 96 Tests
Catalog # ANTIA5834-96
Supplier:  ANTIBODIES.COM
New Product
Rat alpha Skeletal Muscle Actin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of rat alpha Skeletal Muscle Actin in tissue homogenates, cell lysates, cell culture supernates o 1 * 96 Tests
Catalog # ANTIA5834-96
Supplier:  ANTIBODIES.COM