ContactUsLinkComponent
Mouse HP1 gamma/CBX3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse HP1 gamma/CBX3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 96 Tests
Catalog # ANTIA4870-96
Supplier:  ANTIBODIES.COM
New Product
Mouse HP1 gamma/CBX3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse HP1 gamma/CBX3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 96 Tests
Catalog # ANTIA4870-96
Supplier:  ANTIBODIES.COM