ContactUsLinkComponent
Mouse Surfactant Protein D/SP-D ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Surfactant Protein D/SP-D in serum, plasma, lung lavage fluid, tissue homogenates or oth 1 * 96 Tests
  ANTIA4630-96
New Product
Mouse Surfactant Protein D/SP-D ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Surfactant Protein D/SP-D in serum, plasma, lung lavage fluid, tissue homogenates or oth 1 * 96 Tests
  ANTIA4630-96