ContactUsLinkComponent
Guinea Pig IL-8 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of guinea pig IL-8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Catalog # ANTIA3656-96
Supplier:  ANTIBODIES.COM
New Product
Guinea Pig IL-8 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of guinea pig IL-8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologica 1 * 96 Tests
Catalog # ANTIA3656-96
Supplier:  ANTIBODIES.COM