ContactUsLinkComponent
Bovine IL-13 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine IL-13 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 96 Tests
Catalog # ANTIA4161-96
Anbieter:  ANTIBODIES.COM
New Product
Bovine IL-13 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine IL-13 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological flui 1 * 96 Tests
Catalog # ANTIA4161-96
Anbieter:  ANTIBODIES.COM