ContactUsLinkComponent
Mouse Superoxide Dismutase 3/EC-SOD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of mouse Superoxide Dismutase 3/EC-SOD in serum, plasma, tissue homogenates, cell lysates, ce 1 * 96 Tests
Catalog # ANTIA4015-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Superoxide Dismutase 3/EC-SOD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of mouse Superoxide Dismutase 3/EC-SOD in serum, plasma, tissue homogenates, cell lysates, ce 1 * 96 Tests
Catalog # ANTIA4015-96
Supplier:  ANTIBODIES.COM