ContactUsLinkComponent
Mouse ANGPTL2/ARP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of mouse ANGPTL2/ARP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 48 Tests
Catalog # ANTIA5047-48
Supplier:  ANTIBODIES.COM
New Product
Mouse ANGPTL2/ARP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of mouse ANGPTL2/ARP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 48 Tests
Catalog # ANTIA5047-48
Supplier:  ANTIBODIES.COM