ContactUsLinkComponent
Horse TGF beta 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of horse TGF beta 3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 48 Tests
Catalog # ANTIA4109-48
Supplier:  ANTIBODIES.COM
New Product
Horse TGF beta 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of horse TGF beta 3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 48 Tests
Catalog # ANTIA4109-48
Supplier:  ANTIBODIES.COM