ContactUsLinkComponent
This kit uses click chemistry to conjugate HRP to primary antibodies after biotinylation for sequential, high sensitivity multiplex IHC with tyramide signal amplification (TSA). 1 * 250 Tests
Catalog # ANTIA329040-250
Supplier:  ANTIBODIES.COM
New Product
This kit uses click chemistry to conjugate HRP to primary antibodies after biotinylation for sequential, high sensitivity multiplex IHC with tyramide signal amplification (TSA). 1 * 250 Tests
Catalog # ANTIA329040-250
Supplier:  ANTIBODIES.COM