ContactUsLinkComponent
Clear Polypropylene,TPE Cap,Non-Sterilized,With barcode(Tubes: 96/rack, Caps:800/bag, 4800/cs 1 * 4.800 items
Catalog # JETBFTD110020
New Product
Clear Polypropylene,TPE Cap,Non-Sterilized,With barcode(Tubes: 96/rack, Caps:800/bag, 4800/cs 1 * 4.800 items
Catalog # JETBFTD110020