ContactUsLinkComponent
Verticale Laboratory Autoclave Fast Cooling (C) Water cooling coil - Compressed air must be available for the necessary support pressure Vacuum pump (PV) for pre-vacuum air removal and post-vacuum moisture removal for improved drying Steam generator 1 * 1 items
Catalog # TUTT5075ELVCPVG380
Supplier:  TUTTNAUER
New Product
Verticale Laboratory Autoclave Fast Cooling (C) Water cooling coil - Compressed air must be available for the necessary support pressure Vacuum pump (PV) for pre-vacuum air removal and post-vacuum moisture removal for improved drying Steam generator 1 * 1 items
Catalog # TUTT5075ELVCPVG380
Supplier:  TUTTNAUER