ContactUsLinkComponent
Human Eph Receptor A3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human Eph Receptor A3 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA6856-96
New Product
Human Eph Receptor A3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human Eph Receptor A3 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA6856-96