ContactUsLinkComponent
Human Hepatitis B Virus Pre-S1 Antigen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qualitative determination of human Hepatitis B Virus Pre-S1 Antigen in serum, plasma, tissue homogenates, and other bi 1 * 96 Tests
Catalog # ANTIA75125-96
Supplier:  ANTIBODIES.COM
New Product
Human Hepatitis B Virus Pre-S1 Antigen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qualitative determination of human Hepatitis B Virus Pre-S1 Antigen in serum, plasma, tissue homogenates, and other bi 1 * 96 Tests
Catalog # ANTIA75125-96
Supplier:  ANTIBODIES.COM