ContactUsLinkComponent
Human alpha Smooth Muscle Actin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human alpha Smooth Muscle Actin in tissue homogenates, cell lysates or other biological fluids 1 * 48 Tests
  ANTIA82338-48
New Product
Human alpha Smooth Muscle Actin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human alpha Smooth Muscle Actin in tissue homogenates, cell lysates or other biological fluids 1 * 48 Tests
  ANTIA82338-48