Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Human SSH2 ELISA Kits

Human SSH2 ELISA Kits

Human SSH2 ELISA kit
Catalog # ANTIA7360-48
New Product
Supplier:   Antibodies.com
Human SSH2 ELISA Kits
Catalog # ANTIA7360-48
Supplier:  Antibodies.com
New Product
CAS Number:  
Human SSH2 ELISA Kits

Human SSH2 ELISA kit

CAS Number:  
Price
604.00 SFr.
Log in for availability
1 * 48 Tests
Product Details & Documents

Human SSH2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SSH2 in tissue homogenates or other biological fluids.

Human SSH2 ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SSH2 in tissue homogenates or other biological fluids. An antibody specific for SSH2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SSH2 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-SSH2 Antibody, which binds the captured SSH2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of SSH2 captured in each well. The concentration of SSH2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Specifications

Specifications

  • Description:
    Human SSH2 ELISA kit
  • Primary antibody reactivity:
    Human
  • Sample type:
    Tissue homogenates or other biological fluids
  • Detection range:
    0,312 - 20 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cat. No.:
    ANTIA7360-48
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,113 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    SSH2
  • Detection limit:
    Intra-assay: CV <10%, Inter-assay: CV <12%
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.