ContactUsLinkComponent
Guinea Pig beta Nerve Growth Factor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of guinea pig beta Nerve Growth Factor in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA74100-96
Supplier:  ANTIBODIES.COM
New Product
Guinea Pig beta Nerve Growth Factor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of guinea pig beta Nerve Growth Factor in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA74100-96
Supplier:  ANTIBODIES.COM