ContactUsLinkComponent
Rat Estrogen Related Receptor alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of rat Estrogen Related Receptor alpha in tissue homogenates, cell lysates or other biologica 1 * 48 Tests
Catalog # ANTIA8135-48
Supplier:  Antibodies.com
New Product
Rat Estrogen Related Receptor alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of rat Estrogen Related Receptor alpha in tissue homogenates, cell lysates or other biologica 1 * 48 Tests
Catalog # ANTIA8135-48
Supplier:  Antibodies.com