Rat Advanced Glycation End Products/AGE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Advanced Glycation End Products/AGE in serum, plasma, tissue homogenates, and othe 1 * 96 Tests
Catalog # ANTIA80246-96
Anbieter: ANTIBODIES.COM