ContactUsLinkComponent
Mouse IL-13 Receptor alpha 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of mouse IL-13 Receptor alpha 2 in serum, plasma, tissue homogenates, cell lysates, cell culture sup 1 * 96 Tests
Catalog # ANTIA6259-96
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-13 Receptor alpha 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitati ve determination of mouse IL-13 Receptor alpha 2 in serum, plasma, tissue homogenates, cell lysates, cell culture sup 1 * 96 Tests
Catalog # ANTIA6259-96
Supplier:  ANTIBODIES.COM