ContactUsLinkComponent
Human Cytochrome P450 3A4/CYP3A4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Cytochrome P450 3A4/CYP3A4 in tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA6440-48
Supplier:  ANTIBODIES.COM
New Product
Human Cytochrome P450 3A4/CYP3A4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human Cytochrome P450 3A4/CYP3A4 in tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA6440-48
Supplier:  ANTIBODIES.COM