ContactUsLinkComponent
Human HLA Class II DRB1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human HLA Class II DRB1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8070-48
Supplier:  Antibodies.com
New Product
Human HLA Class II DRB1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of human HLA Class II DRB1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA8070-48
Supplier:  Antibodies.com