ContactUsLinkComponent
Mouse alpha 1 Acid Glycoprotein/AGP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of mouse alpha 1 Acid Glycoprotein/AGP in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA75068-96
Supplier:  Antibodies.com
New Product
Mouse alpha 1 Acid Glycoprotein/AGP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of mouse alpha 1 Acid Glycoprotein/AGP in serum, plasma, tissue homogenates, and other biolog 1 * 96 Tests
Catalog # ANTIA75068-96
Supplier:  Antibodies.com